Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rup2
Entrez Gene ID
619560
Gene Name
urinary protein 2
Gene Aliases
UP-1
Chromosome Location
Chr.8: 41657662 - 41661223 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF368860.1 - 1 - 2 71 84
BC127452.1 - 1 - 2 81 84

View Full Product Details