Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sarnp
Entrez Gene ID
362819
Gene Name
SAP domain containing ribonucleoprotein
Gene Aliases
Cip29, RGD1305692
Chromosome Location
Chr.7: 3246191 - 3300467 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033070.1 NP_001028242.1 10 - 11 747 81
XM_006240781.2 XP_006240843.1 9 - 10 707 81
GenBank mRNA BC100070.1 - 10 - 11 747 81

View Full Product Details