Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl13b
Entrez Gene ID
304037
Gene Name
ADP-ribosylation factor-like 13B
Gene Aliases
-
Chromosome Location
Chr.7: 1122567 - 1188209 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107101.1 NP_001100571.1 3 - 4 680 87
XM_006240716.2 XP_006240778.1 2 - 3 461 87
XM_006240717.2 XP_006240779.1 2 - 3 459 87
XM_006240718.2 XP_006240780.1 2 - 3 459 87

View Full Product Details