The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106918.1 | NP_001100388.1 | 3 - 4 | 417 | 88 |
GenBank mRNA | BC160924.1 | - | - | 257 | 88 |