Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plin3
Entrez Gene ID
316130
Gene Name
perilipin 3
Gene Aliases
M6prbp1, Tip47
Chromosome Location
Chr.9: 10769739 - 10781659 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_236783.6 XP_236783.4 1 - 2 106 91

View Full Product Details