Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Khsrp
Entrez Gene ID
171137
Gene Name
KH-type splicing regulatory protein
Gene Aliases
Marta1
Chromosome Location
Chr.9: 10013792 - 10023670 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133602.1 NP_598286.1 3 - 4 382 63
XM_006244292.2 XP_006244354.1 3 - 4 520 63
GenBank mRNA AF308818.1 - 3 - 4 382 63

View Full Product Details