The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007557.1 | NP_001007558.1 | 20 - 21 | 2704 | 100 |
XM_006244280.2 | XP_006244342.1 | 19 - 20 | 2530 | 100 | |
XM_008766759.1 | XP_008764981.1 | 19 - 20 | 2760 | 100 | |
GenBank mRNA | AY686632.1 | - | 20 - 21 | 2704 | 100 |