Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmac
Entrez Gene ID
363327
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
Gene Aliases
-
Chromosome Location
Chr.9: 10338245 - 10341903 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766765.1 XP_008764987.1 1 - 2 127 61

View Full Product Details