Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stard3nl
Entrez Gene ID
291182
Gene Name
STARD3 N-terminal like
Gene Aliases
-
Chromosome Location
Chr.17: 47662553 - 47696453 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008298.1 NP_001008299.1 2 - 3 335 114
GenBank mRNA BC086352.1 - 2 - 3 335 114
FQ220233.1 - 2 - 3 358 114

View Full Product Details