Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amot
Entrez Gene ID
300289
Gene Name
angiomotin
Gene Aliases
RGD1564027
Chromosome Location
Chr.X: 116751231 - 116810233 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257420.2 XP_006257482.1 2 - 3 500 81
XM_006257421.1 XP_006257483.1 3 - 4 187 81

View Full Product Details