Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il13ra2
Entrez Gene ID
171060
Gene Name
interleukin 13 receptor, alpha 2
Gene Aliases
IL-13R-alpha-2
Chromosome Location
Chr.X: 118443958 - 118514716 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133538.2 NP_598222.2 8 - 9 1008 171
XM_006257427.2 XP_006257489.1 7 - 8 876 171
XM_006257428.2 XP_006257490.1 7 - 8 994 171
XM_006257430.2 XP_006257492.1 6 - 7 807 171
XM_006257431.2 XP_006257493.1 4 - 5 515 171
XM_008773388.1 XP_008771610.1 7 - 8 1148 171
XM_008773389.1 XP_008771611.1 8 - 9 1035 171
XM_008773390.1 XP_008771612.1 4 - 5 609 171
XM_008773391.1 XP_008771613.1 4 - 5 1216 171
GenBank mRNA AF448818.1 - 8 - 9 1008 171

View Full Product Details