Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il13ra2
Entrez Gene ID
171060
Gene Name
interleukin 13 receptor, alpha 2
Gene Aliases
IL-13R-alpha-2
Chromosome Location
Chr.X: 118443958 - 118514716 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773391.1 XP_008771613.1 1 - 2 792 105

View Full Product Details