Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slmo2
Entrez Gene ID
494346
Gene Name
slowmo homolog 2 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.3: 172567092 - 172574382 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009543.1 NP_001009543.1 - 253 64
GenBank mRNA BC060590.1 - - 253 64

View Full Product Details