Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfap2c
Entrez Gene ID
362280
Gene Name
transcription factor AP-2 gamma
Gene Aliases
AP2-gamma, Tcfap2c
Chromosome Location
Chr.3: 170550314 - 170559202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201420.2 NP_958823.2 2 - 3 748 85
XM_006235691.2 XP_006235753.1 2 - 3 717 85
GenBank mRNA BC097260.1 - 2 - 3 748 85

View Full Product Details