Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmp7
Entrez Gene ID
85272
Gene Name
bone morphogenetic protein 7
Gene Aliases
BMP-7
Chromosome Location
Chr.3: 170879977 - 170955820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191856.2 NP_001178785.1 1 - 2 843 67
GenBank mRNA AF100787.1 - 1 - 2 88 67

View Full Product Details