Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pa2g4
Entrez Gene ID
288778
Gene Name
proliferation-associated 2G4
Gene Aliases
Ebp1
Chromosome Location
Chr.7: 2979587 - 2987055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004206.1 NP_001004206.1 7 - 8 752 94
GenBank mRNA BC079095.1 - 7 - 8 752 94
FQ233061.1 - 7 - 8 760 94

View Full Product Details