Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ikzf4
Entrez Gene ID
100361923
Gene Name
IKAROS family zinc finger 4
Gene Aliases
-
Chromosome Location
Chr.7: 3061424 - 3079613 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240732.2 XP_006240794.1 1 - 2 292 141

View Full Product Details