Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmel
Entrez Gene ID
362818
Gene Name
premelanosome protein
Gene Aliases
Si, Silv
Chromosome Location
Chr.7: 3133343 - 3143827 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240779.2 XP_006240841.1 10 - 11 2269 62

View Full Product Details