Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pmel
Entrez Gene ID
362818
Gene Name
premelanosome protein
Gene Aliases
Si, Silv
Chromosome Location
Chr.7: 3133343 - 3143827 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240779.2 XP_006240841.1 8 - 9 1981 76

View Full Product Details