Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abcd1
Entrez Gene ID
363516
Gene Name
ATP-binding cassette, subfamily D (ALD), member 1
Gene Aliases
RGD1562128
Chromosome Location
Chr.X: 157074117 - 157095511 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108821.1 NP_001102291.1 9 - 10 2427 72

View Full Product Details