Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnase1l1
Entrez Gene ID
363522
Gene Name
deoxyribonuclease 1-like 1
Gene Aliases
-
Chromosome Location
Chr.X: 156429531 - 156438066 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014223.2 NP_001014245.2 2 - 3 413 238
XM_006229602.1 XP_006229664.1 2 - 3 468 160
GenBank mRNA BC079074.1 - 2 - 3 369 238
DQ116785.1 - 2 - 3 413 238
FQ232580.1 - 2 - 3 417 160

View Full Product Details