Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Emd
Entrez Gene ID
25437
Gene Name
emerin
Gene Aliases
-
Chromosome Location
Chr.X: 156452847 - 156455858 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC059151.1 - 1 - 2 211 89

View Full Product Details