Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tktl1
Entrez Gene ID
689374
Gene Name
transketolase-like 1
Gene Aliases
-
Chromosome Location
Chr.X: 156507797 - 156540733 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109534.1 NP_001103004.1 11 - 12 1487 114

View Full Product Details