Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcdc2
Entrez Gene ID
291130
Gene Name
doublecortin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.17: 41838201 - 42031265 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106110.2 NP_001099580.2 9 - 10 1367 72
GenBank mRNA FM100065.1 - 3 - 4 477 72

View Full Product Details