Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a4
Entrez Gene ID
679784
Gene Name
solute carrier family 17, member 4
Gene Aliases
-
Chromosome Location
Chr.17: 43452322 - 43472192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271214.1 NP_001258143.1 1 - 2 122 108
XM_008771657.1 XP_008769879.1 1 - 2 127 108
XM_008771658.1 XP_008769880.1 2 - 3 441 108
XM_008771659.1 XP_008769881.1 2 - 3 511 108
XM_008771660.1 XP_008769882.1 1 - 2 127 108

View Full Product Details