The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106111.1 | NP_001099581.1 | 2 - 3 | 499 | 95 |
GenBank mRNA | FQ217526.1 | - | 2 - 3 | 449 | 95 |