The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053347.1 | NP_445799.1 | 6 - 7 | 800 | 123 |
XM_006245849.2 | XP_006245911.1 | 7 - 8 | 917 | 123 | |
XM_006245850.2 | XP_006245912.1 | 8 - 9 | 949 | 123 | |
XM_006245851.1 | XP_006245913.1 | 7 - 8 | 917 | 123 | |
XM_008767443.1 | XP_008765665.1 | 8 - 9 | 970 | 123 | |
XM_008767444.1 | XP_008765666.1 | 8 - 9 | 970 | 123 | |
XM_008767445.1 | XP_008765667.1 | 8 - 9 | 970 | 123 | |
GenBank mRNA | AF240463.1 | - | 6 - 7 | 800 | 123 |
BC081759.1 | - | 6 - 7 | 812 | 123 |