Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam7
Entrez Gene ID
29641
Gene Name
ADAM metallopeptidase domain 7
Gene Aliases
7, ADAM, ADAM 7, EAP, EAP I, EAPI, I
Chromosome Location
Chr.15: 49418946 - 49467174 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020301.1 NP_064697.1 14 - 15 1591 59
GenBank mRNA X66140.1 - 14 - 15 1591 59

View Full Product Details