Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gata4
Entrez Gene ID
54254
Gene Name
GATA binding protein 4
Gene Aliases
-
Chromosome Location
Chr.15: 46386703 - 46432914 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144730.1 NP_653331.1 5 - 6 1557 99
XM_006252189.2 XP_006252251.1 4 - 5 1137 99
GenBank mRNA L22761.1 - 5 - 6 1557 99

View Full Product Details