Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lactb
Entrez Gene ID
300803
Gene Name
lactamase, beta
Gene Aliases
-
Chromosome Location
Chr.8: 72750174 - 72766307 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106833.1 NP_001100303.1 1 - 2 440 117
XM_008766242.1 XP_008764464.1 1 - 2 423 117

View Full Product Details