Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ice2
Entrez Gene ID
691379
Gene Name
interactor of little elongation complex ELL subunit 2
Gene Aliases
Narg2
Chromosome Location
Chr.8: 75624938 - 75666781 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191108.1 NP_001178037.1 4 - 5 517 100
XM_006243402.2 XP_006243464.1 5 - 6 985 100

View Full Product Details