Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mfsd7
Entrez Gene ID
305625
Gene Name
major facilitator superfamily domain containing 7
Gene Aliases
RGD1304688
Chromosome Location
Chr.14: 2311122 - 2319574 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107246.1 NP_001100716.1 8 - 9 1407 143

View Full Product Details