The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013987.1 | NP_001014009.1 | 9 - 10 | 1542 | 83 |
XM_006250547.2 | XP_006250609.1 | 9 - 10 | 1667 | 83 | |
XM_006250548.2 | XP_006250610.2 | 9 - 10 | 1657 | 83 | |
XM_008769928.1 | XP_008768150.1 | 9 - 10 | 1657 | 83 | |
XM_008769929.1 | XP_008768151.1 | 9 - 10 | 1657 | 83 | |
XM_008769930.1 | XP_008768152.1 | 7 - 8 | 1328 | 83 | |
GenBank mRNA | BC088426.1 | - | 9 - 10 | 1542 | 83 |