Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim39
Entrez Gene ID
309591
Gene Name
tripartite motif-containing 39
Gene Aliases
Rnf23
Chromosome Location
Chr.20: 2500691 - 2514546 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213562.1 NP_998727.1 6 - 7 1034 117

View Full Product Details