The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009176.2 | NP_001009176.1 | 6 - 7 | 1288 | 116 |
GenBank mRNA | BC129078.1 | - | 6 - 7 | 1288 | 116 |