The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212542.3 | NP_997707.2 | 1 - 2 | 277 | 87 |
GenBank mRNA | BC079252.1 | - | 1 - 2 | 246 | 87 |
EX488885.1 | - | 1 - 2 | 271 | 87 | |
FM088354.1 | - | 1 - 2 | 276 | 87 |