Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mt3
Entrez Gene ID
117038
Gene Name
metallothionein 3
Gene Aliases
GIF, Mt-3
Chromosome Location
Chr.19: 11324708 - 11326112 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053968.3 NP_446420.1 1 - 2 93 81
GenBank mRNA BC058453.1 - 1 - 2 96 81
S65838.1 - 1 - 2 91 81

View Full Product Details