Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sfta2
Entrez Gene ID
415052
Gene Name
surfactant associated 2
Gene Aliases
E030032D13Rik
Chromosome Location
Chr.20: 3604918 - 3605643 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166020.1 NP_001159492.1 2 - 3 167 72

View Full Product Details