Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fignl2
Entrez Gene ID
300238
Gene Name
fidgetin-like 2
Gene Aliases
RGD1566321
Chromosome Location
Chr.7: 142693181 - 142714808 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242373.2 XP_006242435.1 2 - 2 5454 83

View Full Product Details