The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012223.1 | NP_001012223.1 | 4 - 5 | 347 | 85 |
XM_008765773.1 | XP_008763995.1 | 4 - 5 | 374 | 85 | |
GenBank mRNA | BC079406.1 | - | 4 - 5 | 347 | 85 |