Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Faim2
Entrez Gene ID
246274
Gene Name
Fas apoptotic inhibitory molecule 2
Gene Aliases
Lfg, NMP35
Chromosome Location
Chr.7: 141159749 - 141185567 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144756.2 NP_653357.1 5 - 6 513 72
GenBank mRNA AF044201.1 - 5 - 6 462 72
BC087606.1 - 5 - 6 513 72

Gene Symbol
LOC102551901
Entrez Gene ID
102551901
Gene Name
protein lifeguard 2-like
Gene Aliases
-
Chromosome Location
Chr.7: 139047289 - 139063907 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242293.2 XP_006242355.1 5 - 6 739 72

View Full Product Details