Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfcp2
Entrez Gene ID
315309
Gene Name
transcription factor CP2
Gene Aliases
Tcfcp2
Chromosome Location
Chr.7: 142138843 - 142181034 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134714.1 NP_001128186.1 13 - 14 1735 129
XM_006242341.2 XP_006242403.1 14 - 15 1588 129
GenBank mRNA BC168176.1 - 13 - 14 1735 129

View Full Product Details