Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pou6f1
Entrez Gene ID
116545
Gene Name
POU class 6 homeobox 1
Gene Aliases
Brn5
Chromosome Location
Chr.7: 142201614 - 142225256 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105746.1 NP_001099216.1 1 - 2 844 92
XM_006242298.2 XP_006242360.1 7 - 8 1507 92
XM_006242301.2 XP_006242363.1 6 - 7 1744 92
XM_008765654.1 XP_008763876.1 6 - 7 1858 92
XM_008765655.1 XP_008763877.1 7 - 8 1519 92
XM_008765656.1 XP_008763878.1 7 - 8 1577 92
XM_008765657.1 XP_008763879.1 6 - 7 1855 92
XM_008765658.1 XP_008763880.1 6 - 7 1846 92
GenBank mRNA L23204.1 - 2 - 3 821 92

View Full Product Details