Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcrs1
Entrez Gene ID
300222
Gene Name
microspherule protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 140910420 - 140919234 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013106.1 NP_001013124.1 14 - 15 1481 66
GenBank mRNA BC091366.1 - 14 - 15 1481 66

View Full Product Details