Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcrs1
Entrez Gene ID
300222
Gene Name
microspherule protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 140910420 - 140919234 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013106.1 NP_001013124.1 8 - 9 985 93
GenBank mRNA BC091366.1 - 8 - 9 985 93

View Full Product Details