Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxc9
Entrez Gene ID
368178
Gene Name
homeobox C9
Gene Aliases
Hoxc8
Chromosome Location
Chr.7: 144592470 - 144598807 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100907.1 NP_001094377.1 1 - 2 540 94
XM_006242434.2 XP_006242496.1 1 - 2 722 94
GenBank mRNA BQ205031.1 - - 94

View Full Product Details