Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cers5
Entrez Gene ID
366984
Gene Name
ceramide synthase 5
Gene Aliases
Lass5
Chromosome Location
Chr.7: 141386388 - 141424374 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108993.1 NP_001102463.1 7 - 8 900 81
XM_006257374.2 XP_006257436.1 7 - 8 883 81
XM_006257375.2 XP_006257437.1 5 - 6 742 81
GenBank mRNA BC166794.1 - 7 - 8 782 81

View Full Product Details