The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106794.2 | NP_001100264.2 | 5 - 6 | 433 | 66 |
GenBank mRNA | AI576681.1 | - | 1 - 2 | 66 | |
BC169128.1 | - | 5 - 6 | 399 | 66 | |
CF976389.1 | - | 5 - 6 | 437 | 66 |