Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpap3
Entrez Gene ID
300189
Gene Name
RNA polymerase II associated protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 139167302 - 139196355 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004243.1 NP_001004243.1 2 - 3 312 194
XM_008765690.1 XP_008763912.1 3 - 4 656 194
GenBank mRNA BC079414.1 - 2 - 3 312 194

View Full Product Details