Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prph
Entrez Gene ID
24688
Gene Name
peripherin
Gene Aliases
Perf, Prph1
Chromosome Location
Chr.7: 140741633 - 140746197 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012633.2 NP_036765.2 6 - 7 1249 102
XM_006257321.2 XP_006257383.1 5 - 6 2128 102
XM_006257322.2 XP_006257384.1 6 - 7 2030 102
XM_008765669.1 XP_008763891.1 5 - 6 2130 102
GenBank mRNA AF031878.1 - 6 - 7 1242 102
BC100656.1 - 6 - 7 1249 102

View Full Product Details